Merck
  • The Link between ENSO-like Forcing and Hydroclimate Variability of Coastal East Asia during the Last Millennium.
The Link between ENSO-like Forcing and Hydroclimate Variability of Coastal East Asia during the Last Millennium.
Scientific reports (2017-08-16)
Jungjae Park, Jiwoo Han, Qiuhong Jin, Junbeom Bahk, Sangheon Yi
ABSTRACT
Inconsistent reconstructions of East Asian hydroclimate for the last millennium significantly limit our understanding of the mechanisms behind climate variability during the medieval climate anomaly (MCA) and little ice age (LIA) in the region. In this study, we present new high-resolution multiproxy records (diatom, δ