Merck
Alkaline earth metal vanadates as sodium-ion battery anodes.
Nature communications (2017-09-08)
Xiaoming Xu, Chaojiang Niu, Manyi Duan, Xuanpeng Wang, Lei Huang, Junhui Wang, Liting Pu, Wenhao Ren, Changwei Shi, Jiasheng Meng, Bo Song, Liqiang Mai
ABSTRACT
The abundance of sodium resources indicates the potential of sodium-ion batteries as emerging energy storage devices. However, the practical application of sodium-ion batteries is hindered by the limited electrochemical performance of electrode materials, especially at the anode side. Here, we identify alkaline earth metal vanadates as promising anodes for sodium-ion batteries. The prepared calcium vanadate nanowires possess intrinsically high electronic conductivity (> 100 S cm
Sigma-Aldrich
Fluoroethylene carbonate, 99%