Merck
  • Direct Inhibitory Effects of Carbon Monoxide on Six Venoms Containing Fibrinogenolytic Metalloproteinases.
Direct Inhibitory Effects of Carbon Monoxide on Six Venoms Containing Fibrinogenolytic Metalloproteinases.
Basic & clinical pharmacology & toxicology (2016-08-23)
Vance G Nielsen, Philip A Losada
ABSTRACT
Since the introduction of antivenom administration over a century ago to treat venomous snake bite, it has been the most effective therapy for saving life and limb. However, this treatment is not always effective and not without potential life-threatening side effects. We tested a new paradigm to abrogate the plasmatic anticoagulant effects of fibrinogenolytic snake venom metalloproteinases (SVMP) by inhibiting these Zn