Merck
  • Hepatic hypoxia-inducible factors inhibit PPARα expression to exacerbate acetaminophen induced oxidative stress and hepatotoxicity.
Hepatic hypoxia-inducible factors inhibit PPARα expression to exacerbate acetaminophen induced oxidative stress and hepatotoxicity.
Free radical biology & medicine (2017-06-07)
Dawei Li, Yingdong Du, Xiaodong Yuan, Xiaoxiao Han, Zhen Dong, Xiaosong Chen, Haoyu Wu, Jianjun Zhang, Longmei Xu, Conghui Han, Ming Zhang, Qiang Xia
ABSTRACT
Oxidative stress has a critical role in the pathogenesis of acetaminophen (APAP) induced hepatocellular necrosis, and the identification of novel approaches to attenuate oxidative stress is essential to prevent/revert the disease. This study investigated the role of both HIF-1 and HIF-2 in the pathogenesis of APAP-induced oxidative stress, as well as the underlying mechanisms. In the present study, we initially found that knockout of HIF-1α or HIF-2α reduced APAP toxicity, and double knockout afforded the best protection. APAP treatment led to stabilization of both HIF-1α and HIF-2α in mouse livers. Moreover, the protective effects of HIF deficiency were related to the attenuated oxidative stress. Further experiments proved that PPARα, a master regulator in cellular metabolism accounted for the HIF deficiency-caused protective impact on APAP toxicity. Inactivation of HIFs promoted the expression of peroxisome proliferator-activated receptor α (PPARα) in the liver, which in turn activated nuclear factor erythroid 2-related factor 2 (Nrf2). Knockdown of PPARα or Nrf2 negated the hepatoprotection afforded by HIF deficiency. At last,examination of the PPARα promoter identified a HIF-binding site and HIF-dependent repression of PPARα in hepatocytes by luciferase reporter and EMSA study. Taken together, Our results demonstrate that HIFs are key suppressors of PPARα in the liver, thereby compromising the adaptive defense mechanisms against oxidative stress when confronted with APAP. These findings are important to the etiology and therapeutics of APAP hepatotoxicity. The functional link between HIFs and PPARα may have more implications in liver physiology and other pathologic conditions than APAP injury.