Merck
Protein
The Journal of biological chemistry (2016-12-10)
Stephanie M Halmo, Danish Singh, Sneha Patel, Shuo Wang, Melanie Edlin, Geert-Jan Boons, Kelley W Moremen, David Live, Lance Wells
ABSTRACT
Disruption of the