Merck
Selegiline Ameliorates Depression-Like Behavior in Mice Lacking the
Frontiers in behavioral neuroscience (2017-05-19)
Satoka Kasai, Toru Yoshihara, Olga Lopatina, Katsuhiko Ishihara, Haruhiro Higashida
ABSTRACT
Parkinson's disease (PD), a neurodegenerative disorder, is accompanied by various non-motor symptoms including depression and anxiety, which may precede the onset of motor symptoms. Selegiline is an irreversible monoamine oxidase-B (MAO-B) inhibitor, and is widely used in the treatment of PD and major depression. However, there are few reports about the effects of selegiline on non-motor symptoms in PD. The aim of this study was to explore the antidepressant and anxiolytic effects of selegiline, using