- SHAPE analysis of the
SHAPE analysis of the
RNA (New York, N.Y.) (2017-07-26)
Edric K Choi, Kelsey A Ulanowicz, Yen Anh H Nguyen, Jane K Frandsen, Rachel M Mitton-Fry
PMID28739676
ABSTRACT
RNA thermometers regulate expression of some genes involved in virulence of pathogenic bacteria such as