Merck
Thrombospondin-4 mediates TGF-β-induced angiogenesis.
Oncogene (2017-05-10)
S Muppala, R Xiao, I Krukovets, D Verbovetsky, R Yendamuri, N Habib, P Raman, E Plow, O Stenina-Adognravi
ABSTRACT
TGF-β is a multifunctional cytokine affecting many cell types and implicated in tissue remodeling processes. Due to its many functions and cell-specific effects, the consequences of TGF-β signaling are process-and stage-dependent, and it is not uncommon that TGF-β exerts distinct and sometimes opposing effects on a disease progression depending on the stage and on the pathological changes associated with the stage. The mechanisms underlying cell- and process-specific effects of TGF-β are poorly understood. We are describing a novel pathway that mediates induction of angiogenesis in response to TGF-β1. We found that in endothelial cells (EC) thrombospondin-4 (TSP-4), a secreted extracellular matrix (ECM) protein, is upregulated in response to TGF-β1 and mediates the effects of TGF-β1 on angiogenesis. Upregulation of TSP-4 does not require the synthesis of new protein, is not caused by decreased secretion of TSP-4, and is mediated by activation of SMAD3. Using Thbs4
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)