Merck
  • Selection and characterization of alanine racemase inhibitors against Aeromonas hydrophila.
Selection and characterization of alanine racemase inhibitors against Aeromonas hydrophila.
BMC microbiology (2017-05-27)
Yaping Wang, Chao Yang, Wen Xue, Ting Zhang, Xipei Liu, Jiansong Ju, Baohua Zhao, Dong Liu
ABSTRACT
Combining experimental and computational screening methods has been of keen interest in drug discovery. In the present study, we developed an efficient screening method that has been used to screen 2100 small-molecule compounds for alanine racemase Alr-2 inhibitors. We identified ten novel non-substrate Alr-2 inhibitors, of which patulin, homogentisic acid, and hydroquinone were active against Aeromonas hydrophila. The compounds were found to be capable of inhibiting Alr-2 to different extents with 50% inhibitory concentrations (IC Our findings suggested that combining experimental and computational methods could be used for an efficient, large-scale screening of alanine racemase inhibitors against A. hydrophila that could be applied in the development of new antibiotics against A. hydrophila.