- Comprehensive Analysis of Codon Usage Bias in Seven
Comprehensive Analysis of Codon Usage Bias in Seven
Frontiers in microbiology (2017-08-12)
Hui Song, Jing Liu, Qiuyan Song, Qingping Zhang, Pei Tian, Zhibiao Nan
PMID28798739
ABSTRACT
Codon usage bias plays an important role in shaping genomes and genes in unicellular species and multicellular species. Here, we first analyzed codon usage bias in seven