- A systematic study of the influence of mesoscale structuring on the kinetics of a chemical reaction.
A systematic study of the influence of mesoscale structuring on the kinetics of a chemical reaction.
Physical chemistry chemical physics : PCCP (2017-07-01)
Sebastian Krickl, Thomas Buchecker, Andreas Uwe Meyer, Isabelle Grillo, Didier Touraud, Pierre Bauduin, Burkhard König, Arno Pfitzner, Werner Kunz
PMID28665425
ABSTRACT
In this contribution, we (i) link the mesoscopic structuring of the binary structured solvent mixture H