Merck
Atmospheric deposition, CO
Scientific reports (2017-08-31)
M Fernández-Martínez, S Vicca, I A Janssens, P Ciais, M Obersteiner, M Bartrons, J Sardans, A Verger, J G Canadell, F Chevallier, X Wang, C Bernhofer, P S Curtis, D Gianelle, T Grünwald, B Heinesch, A Ibrom, A Knohl, T Laurila, B E Law, J M Limousin, B Longdoz, D Loustau, I Mammarella, G Matteucci, R K Monson, L Montagnani, E J Moors, J W Munger, D Papale, S L Piao, J Peñuelas
ABSTRACT
Concentrations of atmospheric carbon dioxide (CO