- Temporary impact of blood donation on physical performance and hematologic variables in women.
Temporary impact of blood donation on physical performance and hematologic variables in women.
Transfusion (2017-04-30)
Ida Stangerup, Nana L Kramp, Andreas K Ziegler, Flemming Dela, Karin Magnussen, Jørn Wulff Helge
PMID28452395
ABSTRACT
No former studies have examined how blood donation influences physical performance in women, who due to menstruation may have a slower recovery of performance. Therefore, the aim of this study was to clarify how VO VO VO VO