Merck
Unusual infrared absorption increases in photo-degraded organic films.
Nanoscale (2017-06-15)
Satvik Shah, Rana Biswas, Thomas Koschny, Vikram Dalal
ABSTRACT
Degradation is among the most pressing problems facing organic materials, occurring through ingress of moisture and oxygen, and light exposure. We determine the nanoscale pathways underlying degradation by light-soaking organic films in an environmental chamber, and performing infrared spectroscopy, to identify atomic bonding changes. We utilize as a prototype the low band gap PTB7-PCBM blend. Films light-soaked in the presence of oxygen show unusual increased absorption at 1727 cm