Merck
  • CD73 on T Cells Orchestrates Cardiac Wound Healing After Myocardial Infarction by Purinergic Metabolic Reprogramming.
CD73 on T Cells Orchestrates Cardiac Wound Healing After Myocardial Infarction by Purinergic Metabolic Reprogramming.
Circulation (2017-04-23)
Nadine Borg, Christina Alter, Nicole Görldt, Christoph Jacoby, Zhaoping Ding, Bodo Steckel, Christine Quast, Florian Bönner, Daniela Friebe, Sebastian Temme, Ulrich Flögel, Jürgen Schrader
ABSTRACT
T cells are required for proper healing after myocardial infarction. The mechanism of their beneficial action, however, is unknown. The proinflammatory danger signal ATP, released from damaged cells, is degraded by the ectonucleotidases CD39 and CD73 to the anti-inflammatory mediator adenosine. Here, we investigate the contribution of CD73-derived adenosine produced by T cells to cardiac remodeling after ischemia/reperfusion and define its mechanism of action. Myocardial ischemia (50 minutes followed by reperfusion) was induced in global CD73 Changes in functional parameters of CD4-CD73 This work demonstrates that CD73 on T cells plays a crucial role in the cardiac wound healing process after myocardial infarction. The underlying mechanism involves a profound increase in the hydrolysis of ATP/NAD and AMP, resulting primarily from the upregulation of pyrophosphatases and CD73. We also define A