Merck
  • Inhibition of Methotrexate Uptake via Organic Anion Transporters OAT1 and OAT3 by Glucuronides of Nonsteroidal Anti-inflammatory Drugs.
Inhibition of Methotrexate Uptake via Organic Anion Transporters OAT1 and OAT3 by Glucuronides of Nonsteroidal Anti-inflammatory Drugs.
Biological & pharmaceutical bulletin (2017-06-02)
Masahiro Iwaki, Hiroaki Shimada, Yuri Irino, Manami Take, Sachiko Egashira
ABSTRACT
Combination therapy of non-steroidal anti-inflammatory drugs (NSAIDs) and methotrexate (MTX) sometimes triggers adverse effects, such as liver injury, renal failure, gastrointestinal disorders, and myelosuppression, owing to the reduction of MTX clearance. Previous reports have suggested that NSAIDs inhibit renal MTX uptake via organic anion transporters (OATs) and reduced folate transporter (RFC)-1 and efflux via multidrug resistance-associated proteins (MRPs). Recently, our laboratory found inhibitory effects of NSAIDs-glucuronide (NSAIDs-Glu), a major metabolite of NSAIDs, on MRP-mediated MTX transport as a new site of interaction between MTX and NSAIDs. However, it remains unclear that whether NSAIDs-Glu inhibit renal uptake of MTX. Therefore, the present study aimed to evaluate inhibitory effects of several NSAIDs-Glu (diclofenac, R- and S-ibuprofen, R- and S-flurbiprofen, and R- and S-naproxen) on human OAT1 and OAT3-mediated MTX transport. In this study, [