Merck
  • CD200R1 agonist attenuates mechanisms of chronic disease in a murine model of multiple sclerosis.
CD200R1 agonist attenuates mechanisms of chronic disease in a murine model of multiple sclerosis.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2010-02-12)
Yingru Liu, Yoshio Bando, David Vargas-Lowy, Wassim Elyaman, Samia J Khoury, Tao Huang, Karin Reif, Tanuja Chitnis
ABSTRACT
To assess the effects and mechanisms of a CD200R1 agonist administered during the progressive stage of a multiple sclerosis model, we administered CD200R1 agonist (CD200Fc) or control IgG2a during the chronic phase of disease (days 10-30) in mice with experimental autoimmune encephalomyelitis (EAE), induced using myelin oligodendrocyte glycoprotein peptide 35-55 (MOG35-55) peptide. We found that administration of CD200Fc during the chronic stages of EAE reduced disease severity, demyelination, and axonal damage, through the modulation of several key disease mechanisms. CD200Fc treatment suppressed macrophage and microglial accumulation within the CNS, in part through downregulation of adhesion molecules VLA-4 and LFA-1, which are necessary for macrophage migration. Additionally, expression of activation markers MHC-II and CD80 and production of proinflammatory cytokines IL-6, tumor necrosis factor-alpha, and nitric oxide by CD11b(+) cells were decreased in both the spleen and CNS in CD200Fc-treated animals. Antigen-presenting cell function in the spleen and CNS was suppressed in CD200Fc-treated mice, but there were no significant alterations on T cell activation or phenotype. CD200Fc increased apoptosis of CD11b(+) cells but not astrocytes. In contrast, addition of CD200Fc treatment protected oligodendrocytes from apoptosis in vitro and in vivo. Our results demonstrate that CD200R1 agonists modulate both myeloid- and non-myeloid-related mechanisms of chronic disease in the EAE model and may be effective in the treatment of progressive multiple sclerosis and other neurodegenerative diseases.