Merck
  • APOBEC3B and IL-6 form a positive feedback loop in hepatocellular carcinoma cells.
APOBEC3B and IL-6 form a positive feedback loop in hepatocellular carcinoma cells.
Science China. Life sciences (2017-06-25)
Shuran Li, Xueyang Bao, Duowei Wang, Linjun You, Xianjing Li, Hongbao Yang, Jinsong Bian, Yun Wang, Yong Yang
ABSTRACT
APOBEC3 protein families, a DNA cytidine deaminase, were up-regulated in multiple tumors. However, the relationship between Hepatocellular carcinoma (HCC) and APOBEC3B (A3B) remains unknown. It has been confirmed that interleukin-6 (IL-6) has significant impacts on oncogenesis of HCC. Here, we reported that the expression of IL-6 was substantially up-regulated by A3B in HepG2 cells. A3B induced IL-6 expression through relocating HuR to enhance the IL-6 mRNA stability. Further analysis indicated that IL-6 also increased the expression of A3B through JAK1/STAT3 signaling pathway, which formed a positive feedback to maintain the continuous expression of A3B and IL-6, and thereby promoted the prolonged non-resolving inflammation. Collectively, these findings suggest that A3B is essential for oncogenesis of HCC, and is a potential target for preventive intervention.