Merck
Target Cell Type-Dependent Differences in Ca
The Journal of neuroscience : the official journal of the Society for Neuroscience (2017-01-25)
Tímea Éltes, Tekla Kirizs, Zoltan Nusser, Noemi Holderith
ABSTRACT
Target cell type-dependent differences in presynaptic release probability (