Merck
  • PAX3-FOXO1 Establishes Myogenic Super Enhancers and Confers BET Bromodomain Vulnerability.
PAX3-FOXO1 Establishes Myogenic Super Enhancers and Confers BET Bromodomain Vulnerability.
Cancer discovery (2017-04-28)
Berkley E Gryder, Marielle E Yohe, Hsien-Chao Chou, Xiaohu Zhang, Joana Marques, Marco Wachtel, Beat Schaefer, Nirmalya Sen, Young Song, Alberto Gualtieri, Silvia Pomella, Rossella Rota, Abigail Cleveland, Xinyu Wen, Sivasish Sindiri, Jun S Wei, Frederic G Barr, Sudipto Das, Thorkell Andresson, Rajarshi Guha, Madhu Lal-Nag, Marc Ferrer, Jack F Shern, Keji Zhao, Craig J Thomas, Javed Khan
ABSTRACT
Alveolar rhabdomyosarcoma is a life-threatening myogenic cancer of children and adolescent young adults, driven primarily by the chimeric transcription factor PAX3-FOXO1. The mechanisms by which PAX3-FOXO1 dysregulates chromatin are unknown. We find PAX3-FOXO1 reprograms the