Merck
  • Inclusion of wheat and triticale silage in the diet of lactating dairy cows.
Inclusion of wheat and triticale silage in the diet of lactating dairy cows.
Journal of dairy science (2017-06-12)
M T Harper, J Oh, F Giallongo, G W Roth, A N Hristov
ABSTRACT
The objective of this experiment was to partially replace corn silage with 2 alternative forages, wheat (Triticum aestivum) or triticale (X Triticosecale) silages at 10% of the diet dry matter (DM), and investigate the effects on dairy cow productivity, nutrient utilization, enteric CH