Merck
  • Capecitabine With Mitomycin Reduces Acute Hematologic Toxicity and Treatment Delays in Patients Undergoing Definitive Chemoradiation Using Intensity Modulated Radiation Therapy for Anal Cancer.
Capecitabine With Mitomycin Reduces Acute Hematologic Toxicity and Treatment Delays in Patients Undergoing Definitive Chemoradiation Using Intensity Modulated Radiation Therapy for Anal Cancer.
International journal of radiation oncology, biology, physics (2017-07-20)
Karyn A Goodman, Diana Julie, Andrea Cercek, Lajhem Cambridge, Kaitlin M Woo, Zhigang Zhang, Abraham J Wu, Diane L Reidy, Neil H Segal, Zsofia K Stadler, Leonard B Saltz
ABSTRACT
To assess the impact on acute toxicity of replacing 5-fluorouracil (5-FU) with capecitabine in definitive chemoradiation for patients with anal squamous cell carcinoma (ASCC). We retrospectively reviewed the records of 107 consecutive patients with nonmetastatic ASCC treated with definitive chemoradiation from January 2009 to May 2014. In 2011, based on the noninferiority of capecitabine versus 5-FU, our institutional practice shifted to use capecitabine instead of 5-FU for ASCC. Of 107 patients, 63 were treated with infusional 5-FU (1000 mg/m The median age at diagnosis was 59 years, and 78 patients (73%) were female. The patient characteristics were similar between the 2 treatment groups. All patients in both groups were treated with intensity modulated RT (median dose, 56 Gy). In the 5-FU group, 52% experienced grade 3 to 4 neutropenia compared with 20% in the capecitabine group (P=.001). Treatment breaks resulting from toxicity, primarily related to grade 3+ hematologic toxicity, were necessary for 42% of patients treated with 5-FU versus 16% of those treated with capecitabine (P=.006). Pelvic radiation therapy with MMC plus capecitabine was well tolerated and appeared to have less grade 3+ acute hematologic toxicity and fewer treatment interruptions than in a population of ASCC patients undergoing definitive chemoradiation with MMC and 5-FU.