Merck
  • Expression of Heterologous Sigma Factor Expands the Searchable Space for Biofuel Tolerance Mechanisms.
Expression of Heterologous Sigma Factor Expands the Searchable Space for Biofuel Tolerance Mechanisms.
ACS synthetic biology (2017-03-21)
Timothy A Tomko, Mary J Dunlop
ABSTRACT
Microorganisms can produce hydrocarbons that can serve as replacements or additions to conventional liquid fuels for use in the transportation sector. However, a common problem in the microbial synthesis of biofuels is that these compounds often have toxic effects on the cell. In this study, we focused on mitigating the toxicity of the biojet fuel precursor pinene on Escherichia coli. We used genomic DNA from Pseudomonas putida KT2440, which has innate solvent-tolerance properties, to create transgenic libraries in an E. coli host. We exposed cells containing the library to pinene, selecting for genes that improved tolerance. Importantly, we found that expressing the sigma factor RpoD from P. putida greatly expanded the diversity of tolerance genes recovered. With low expression of rpoD