- Dapoxetine induces neuroprotective effects against glutamate-induced neuronal cell death by inhibiting calcium signaling and mitochondrial depolarization in cultured rat hippocampal neurons.
Dapoxetine induces neuroprotective effects against glutamate-induced neuronal cell death by inhibiting calcium signaling and mitochondrial depolarization in cultured rat hippocampal neurons.
European journal of pharmacology (2017-03-23)
Imju Jeong, Ji Seon Yang, Yi Jae Hong, Hee Jung Kim, Sang June Hahn, Shin Hee Yoon
PMID28322832
ABSTRACT
Selective serotonin reuptake inhibitors (SSRIs) have an inhibitory effect on various ion channels including Ca