Merck
  • Placental Pim-1 expression is increased in obesity and regulates cytokine- and toll-like receptor-mediated inflammation.
Placental Pim-1 expression is increased in obesity and regulates cytokine- and toll-like receptor-mediated inflammation.
Placenta (2017-05-11)
Stella Liong, Gillian Barker, Martha Lappas
ABSTRACT
Obesity is a growing epidemic, and as a consequence the number of obese pregnancies has also increased. Pregnancy is characterised by maternal and placental inflammation which is intensified with maternal obesity. The proviral integration site for Moloney murine leukemic virus (Pim)-1 protein is a serine/threonine kinase involved in a wide range of inflammatory diseases. In relation to obesity, however, its role has not been elucidated in human placenta. The aims were to determine the placental expression of Pim-1 with pre-existing maternal obesity and its role in regulating placental inflammation associated with obesity. Human placenta was obtained at the time of term Caesarean section from lean and pre-existing obese pregnant women to determine the effect of maternal obesity on Pim-1 expression. To determine the effect of Pim-1 on the inflammatory response induced by bacterial endotoxin LPS and pro-inflammatory cytokines TNF-α or IL-1β, the chemical inhibitor SMI-4a and siRNA were used. Pim-1 protein and mRNA expression was significantly increased in placenta of obese women. SMI-4a significantly suppressed the expression and release of pro-inflammatory cytokine IL-6 and chemokines GRO-α and MCP-1 when stimulated with LPS or TNF-α in placenta. Primary trophoblast cells transfected with Pim-1 siRNA had decreased expression and release of pro-inflammatory cytokines IL-1β, IL-6, chemokines GRO-α and MCP-1, when stimulated with LPS, TNF-α or IL-1β. The findings from this study implicate Pim-1 may contribute to placental inflammation in pregnancies complicated by maternal obesity. Thus, therapeutic targets for Pim-1 may improve fetal outcomes complicated by obese pregnancies.