Merck
  • Albumin-Gold Nanorod Nanoplatform for Cell-Mediated Tumoritropic Delivery with Homogenous ChemoDrug Distribution and Enhanced Retention Ability.
Albumin-Gold Nanorod Nanoplatform for Cell-Mediated Tumoritropic Delivery with Homogenous ChemoDrug Distribution and Enhanced Retention Ability.
Theranostics (2017-08-26)
Hsien-Ting Chiu, Cheng-Kuan Su, Yuh-Chang Sun, Chi-Shiun Chiang, Yu-Fen Huang
ABSTRACT
Recently, living cells with tumor-homing properties have provided an exciting opportunity to achieve optimal delivery of nanotherapeutic agents. However, premature payload leakage may impair the host cells, often leading to inadequate