Merck
  • Pterostilbene, a Methoxylated Resveratrol Derivative, Efficiently Eradicates Planktonic, Biofilm, and Intracellular MRSA by Topical Application.
Pterostilbene, a Methoxylated Resveratrol Derivative, Efficiently Eradicates Planktonic, Biofilm, and Intracellular MRSA by Topical Application.
Frontiers in microbiology (2017-07-01)
Shih-Chun Yang, Chih-Hua Tseng, Pei-Wen Wang, Po-Liang Lu, Yi-Han Weng, Feng-Lin Yen, Jia-You Fang
ABSTRACT
Pterostilbene is a methoxylated derivative of resveratrol originated from natural sources. We investigated the antibacterial activity of pterostilbene against drug-resistant