- Enhanced transient receptor potential channel-mediated Ca
Enhanced transient receptor potential channel-mediated Ca
Fundamental & clinical pharmacology (2017-01-21)
Kazuo Murakami, Tomohiro Osanai, Makoto Tanaka, Kimitaka Nishizaki, Takahiko Kinjo, Tomohiro Tanno, Yuji Ishida, Akiko Suzuki, Tomohide Endo, Hirofumi Tomita, Ken Okumura
PMID28107550
ABSTRACT
We reported that coronary spasm was induced in the transgenic mice with the increased phospholipase C (PLC)-δ1 activity. We investigated the effect of enhanced PLC-δ1 on Ca