Merck
  • SLC10A4 regulates IgE-mediated mast cell degranulation in vitro and mast cell-mediated reactions in vivo.
SLC10A4 regulates IgE-mediated mast cell degranulation in vitro and mast cell-mediated reactions in vivo.
Scientific reports (2017-04-26)
Hanna Pettersson, Behdad Zarnegar, Annika Westin, Viktor Persson, Christiane Peuckert, Jörgen Jonsson, Jenny Hallgren, Klas Kullander
ABSTRACT
Mast cells act as sensors in innate immunity and as effector cells in adaptive immune reactions. Here we demonstrate that SLC10A4, also referred to as the vesicular aminergic-associated transporter, VAAT, modifies mast cell degranulation. Strikingly, Slc10a4
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%