Merck
  • Determination of Two Sulfonylurea Herbicides Residues in Soil Environment Using HPLC and Phytotoxicity of These Herbicides by Lentil Bioassay.
Determination of Two Sulfonylurea Herbicides Residues in Soil Environment Using HPLC and Phytotoxicity of These Herbicides by Lentil Bioassay.
Bulletin of environmental contamination and toxicology (2017-04-05)
Mohammad Mehdizadeh, Mohammad Taghi Alebrahim, Mahmoud Roushani
ABSTRACT
A HPLC-UV detection system was used for determination of sulfosulfuron and tribenuron methyl residues from soils. The soils were fortified with sulfosulfuron and tribenuron methyl at rates of 26 and 15 g a.i. ha