Merck
  • The Influence of Land Use Intensity on the Plant-Associated Microbiome of
The Influence of Land Use Intensity on the Plant-Associated Microbiome of
Frontiers in plant science (2017-07-07)
Jennifer Estendorfer, Barbara Stempfhuber, Paula Haury, Gisle Vestergaard, Matthias C Rillig, Jasmin Joshi, Peter Schröder, Michael Schloter
ABSTRACT
In this study, we investigated the impact of different land use intensities (LUI) on the root-associated microbiome of