Merck
  • Role of cyclooxygenase-mediated metabolites in lipid metabolism and expression of some immune-related genes in juvenile grass carp (Ctenopharyngodon idellus) fed arachidonic acid.
Role of cyclooxygenase-mediated metabolites in lipid metabolism and expression of some immune-related genes in juvenile grass carp (Ctenopharyngodon idellus) fed arachidonic acid.
Fish physiology and biochemistry (2016-12-25)
Jing-Jing Tian, Cai-Xia Lei, Hong Ji, Ai Jin
ABSTRACT
Cyclooxygenase (COX) catalyzes the conversion of arachidonic acid (ARA) to prostaglandins, and COX-mediated metabolites play important roles in the regulation of lipid metabolism and immunity in mammals. However, such roles of COX in fish remain largely unknown. In this study, we designed three semi-purified diets, namely ARA-free (control), ARA, and ARA + acetylsalicylic acid (ASA; a COX inhibitor), and used them to feed grass carp (27.65 ± 3.05 g) for 8 weeks. The results showed that dietary ARA significantly increased the amount of ARA in the hepatopancreas, muscle, and kidney (P < 0.05), whereas this increase was reduced by dietary ASA. The hepatopancreatic prostaglandin E