- Lipocalin-2 Promotes Pancreatic Ductal Adenocarcinoma by Regulating Inflammation in the Tumor Microenvironment.
Lipocalin-2 Promotes Pancreatic Ductal Adenocarcinoma by Regulating Inflammation in the Tumor Microenvironment.
Cancer research (2017-03-03)
Sobeyda B Gomez-Chou, Agnieszka Katarzyna Swidnicka-Siergiejko, Niharika Badi, Myrriah Chavez-Tomar, Gregory B Lesinski, Tanios Bekaii-Saab, Matthew R Farren, Thomas A Mace, Carl Schmidt, Yan Liu, Defeng Deng, Rosa F Hwang, Liran Zhou, Todd Moore, Deyali Chatterjee, Huamin Wang, Xiaohong Leng, Ralph B Arlinghaus, Craig D Logsdon, Zobeida Cruz-Monserrate
PMID28249896
ABSTRACT
Lipocalin-2 (LCN2) promotes malignant development in many cancer types. LCN2 is upregulated in patients with pancreatic ductal adenocarcinoma (PDAC) and in obese individuals, but whether it contributes to PDAC development is unclear. In this study, we investigated the effects of Lcn2 depletion on diet-induced obesity, inflammation, and PDAC development. Mice with acinar cell-specific expression of Kras