Merck
  • Two-step conversion of carboxylic esters into distally fluorinated ketones via ring cleavage of cyclopropanol intermediates: application of sulfinate salts as fluoroalkylating reagents.
Two-step conversion of carboxylic esters into distally fluorinated ketones via ring cleavage of cyclopropanol intermediates: application of sulfinate salts as fluoroalkylating reagents.
Organic & biomolecular chemistry (2017-05-18)
Yulia A Konik, Marina Kudrjashova, Nele Konrad, Sandra Kaabel, Ivar Järving, Margus Lopp, Dzmitry G Kananovich
ABSTRACT
Tertiary cyclopropanols easily available from carboxylic esters have been used in the synthesis of distally fluorinated ketones. Cyclopropane ring cleavage reactions in methanol with aqueous tert-butyl hydroperoxide in the presence of a copper(ii) acetate catalyst and sodium triflinate (Langlois reagent) afford β-trifluoromethyl ketones in 16-74% isolated yields. Sodium triflinate serves as a precursor of reactive trifluoromethyl copper species, enabling ring-opening trifluoromethylation, as evidenced by mechanistic studies. We also demonstrate here that other sulfinate salts, such as sodium 1,1-difluoroethanesulfinate, sodium 2-(4-bromophenyl)-1,1-difluoroethanesulfinate and sodium 1-(trifluoromethyl)cyclopropanesulfinate, can be used as fluoroalkylation reagents, resulting in the corresponding fluorinated ketones.