Merck
  • Active shortening protects against stretch-induced force deficits in human skeletal muscle.
Active shortening protects against stretch-induced force deficits in human skeletal muscle.
Journal of applied physiology (Bethesda, Md. : 1985) (2017-02-27)
Anjali L Saripalli, Kristoffer B Sugg, Christopher L Mendias, Susan V Brooks, Dennis R Claflin
ABSTRACT
Skeletal muscle contraction results from molecular interactions of myosin "crossbridges" with adjacent actin filament binding sites. The binding of myosin to actin can be "weak" or "strong," and only strong binding states contribute to force production. During active shortening, the number of strongly bound crossbridges declines with increasing shortening velocity. Forcibly stretching a muscle that is actively shortening at high velocity results in no apparent negative consequences, whereas stretch of an isometrically (fixed-length) contracting muscle causes ultrastructural damage and a decline in force-generating capability. Our working hypothesis is that stretch-induced damage is uniquely attributable to the population of crossbridges that are strongly bound. We tested the hypothesis that stretch-induced force deficits decline as the prevailing shortening velocity is increased. Experiments were performed on permeabilized segments of individual skeletal muscle fibers obtained from human subjects. Fibers were maximally activated and allowed either to generate maximum isometric force (F