Merck
  • Possible involvement of the CA1 GABAergic system on harmaline induced memory consolidation deficit.
Possible involvement of the CA1 GABAergic system on harmaline induced memory consolidation deficit.
Brain research bulletin (2017-01-24)
Mohammad Nasehi, Naghmeh Saadati, Fatemeh Khakpai, Mohammad-Reza Zarrindast
ABSTRACT
Activation of the GABA