- Regulation of Strigolactone Biosynthesis by Gibberellin Signaling.
Regulation of Strigolactone Biosynthesis by Gibberellin Signaling.
Plant physiology (2017-04-14)
Shinsaku Ito, Daichi Yamagami, Mikihisa Umehara, Atsushi Hanada, Satoko Yoshida, Yasuyuki Sasaki, Shunsuke Yajima, Junko Kyozuka, Miyako Ueguchi-Tanaka, Makoto Matsuoka, Ken Shirasu, Shinjiro Yamaguchi, Tadao Asami
PMID28404726
ABSTRACT
Strigolactones (SLs) are a class of plant hormones that regulate diverse physiological processes, including shoot branching and root development. They also act as rhizosphere signaling molecules to stimulate the germination of root parasitic weeds and the branching of arbuscular mycorrhizal fungi. Although various types of cross talk between SLs and other hormones have been reported in physiological analyses, the cross talk between gibberellin (GA) and SLs is poorly understood. We screened for chemicals that regulate the level of SLs in rice (