- Contribution of platelet P2Y
Contribution of platelet P2Y
Journal of thrombosis and haemostasis : JTH (2017-03-28)
K Bekő, B Koványi, F Gölöncsér, G Horváth, Á Dénes, Z Környei, B Botz, Z Helyes, C E Müller, B Sperlágh
PMID28345287
ABSTRACT
Essentials The role of platelet P2Y Background P2Y