Merck
  • Realizing Full Coverage of Stable Perovskite Film by Modified Anti-Solvent Process.
Realizing Full Coverage of Stable Perovskite Film by Modified Anti-Solvent Process.
Nanoscale research letters (2017-05-26)
Long Ji, Ting Zhang, Yafei Wang, Peng Zhang, Detao Liu, Zhi Chen, Shibin Li
ABSTRACT
Lead-free solution-processed solid-state photovoltaic devices based on formamidinium tin triiodide (FASnI