Merck
  • High-Efficiency Light-Emitting Diodes of Organometal Halide Perovskite Amorphous Nanoparticles.
High-Efficiency Light-Emitting Diodes of Organometal Halide Perovskite Amorphous Nanoparticles.
ACS nano (2016-06-11)
Jun Xing, Fei Yan, Yawen Zhao, Shi Chen, Huakang Yu, Qing Zhang, Rongguang Zeng, Hilmi Volkan Demir, Xiaowei Sun, Alfred Huan, Qihua Xiong
ABSTRACT
Organometal halide perovskite has recently emerged as a very promising family of materials with augmented performance in electronic and optoelectronic applications including photovoltaic devices, photodetectors, and light-emitting diodes. Herein, we propose and demonstrate facile solution synthesis of a series of colloidal organometal halide perovskite CH3NH3PbX3 (X = halides) nanoparticles with amorphous structure, which exhibit high quantum yield and tunable emission from ultraviolet to near-infrared. The growth mechanism and photoluminescence properties of the perovskite amorphous nanoparticles were studied in detail. A high-efficiency green-light-emitting diode based on amorphous CH3NH3PbBr3 nanoparticles was demonstrated. The perovskite amorphous nanoparticle-based light-emitting diode shows a maximum luminous efficiency of 11.49 cd/A, a power efficiency of 7.84 lm/W, and an external quantum efficiency of 3.8%, which is 3.5 times higher than that of the best colloidal perovskite quantum-dot-based light-emitting diodes previously reported. Our findings indicate the great potential of colloidal perovskite amorphous nanoparticles in light-emitting devices.