- Life on the edge: O2 binding in Atlantic cod red blood cells near their southern distribution limit is not sensitive to temperature or haemoglobin genotype.
Life on the edge: O2 binding in Atlantic cod red blood cells near their southern distribution limit is not sensitive to temperature or haemoglobin genotype.
The Journal of experimental biology (2017-02-06)
Samantha L Barlow, Julian Metcalfe, David A Righton, Michael Berenbrink
PMID28148818
ABSTRACT
Atlantic cod are a commercially important species believed to be threatened by warming seas near their southern, equatorward upper thermal edge of distribution. Limitations to circulatory O