Merck
  • Decreased expression of semaphorin 3D is associated with genesis and development in colorectal cancer.
Decreased expression of semaphorin 3D is associated with genesis and development in colorectal cancer.
World journal of surgical oncology (2017-03-23)
Zhen Wang, Meiman Ding, Naiying Qian, Beifeng Song, Jiayin Yu, Jinlong Tang, Jingyu Wang
ABSTRACT
Semaphorin 3D (SEMA3D) plays important roles in the genesis and progress of many cancers. However, the relationship between SEMA3D and colorectal cancer (CRC) remains unknown. The aim of this study was to investigate whether SEMA3D can be used as a predictive marker for the diagnosis, metastasis, and prognosis of CRC by assessing the expression of SEMA3D in the tissues and serum of CRC patients. Real-time quantitative polymerase chain reaction (qPCR) was used to measure the expression of SEMA3D mRNA in 100 CRC tissues and matched normal tissues. qPCR was also used to detect the expression of SEMA3D mRNA in the CRC cell line RKO. RKO cells were transfected with SEMA3D small-interring RNA (siRNA) to interfere with endogenous SEMA3D. The migratory ability of control and SEMA3D siRNA-transfected RKO cells was determined by transwell assays. Enzyme-linked immunosorbent assay (ELISA) was utilized to detect the levels of SEMA3D in the serum of 80 CRC patients and 100 normal healthy controls. The expression of SEMA3D in 215 CRC tissues was assessed using immunohistochemistry (IHC). Then, statistical analyses were adopted to assess SEMA3D protein levels and clinical pathological characteristics. The mRNA expression of SEMA3D was significantly lower in CRC tissues than in paired normal tissues (t = 5.027, P < 0.0001). Compared with normal healthy controls, the serum levels of SEMA3D were decreased significantly in CRC patients (t = 3.656, P = 0.0003). The expression of SEMA3D protein was linked to lymph node metastasis, and low expression led to lymph node metastasis (χ SEMA3D might function as a tumor suppressor during the formation and development of CRC. SEMA3D might become a predictive marker for the diagnosis, metastasis, and prognosis of CRC and provide a novel target for the prevention and treatment of CRC.