- Conditional knockout of Fgf13 in murine hearts increases arrhythmia susceptibility and reveals novel ion channel modulatory roles.
Conditional knockout of Fgf13 in murine hearts increases arrhythmia susceptibility and reveals novel ion channel modulatory roles.
Journal of molecular and cellular cardiology (2017-01-26)
Xiangchong Wang, He Tang, Eric Q Wei, Zhihua Wang, Jing Yang, Rong Yang, Sheng Wang, Yongjian Zhang, Geoffrey S Pitt, Hailin Zhang, Chuan Wang
PMID28119060
ABSTRACT
The intracellular fibroblast growth factors (iFGF/FHFs) bind directly to cardiac voltage gated Na