Merck
  • Physiological and Molecular Characterization of Hydroxyphenylpyruvate Dioxygenase (HPPD)-inhibitor Resistance in Palmer Amaranth (
Physiological and Molecular Characterization of Hydroxyphenylpyruvate Dioxygenase (HPPD)-inhibitor Resistance in Palmer Amaranth (
Frontiers in plant science (2017-04-27)
Sridevi Nakka, Amar S Godar, Prashant S Wani, Curtis R Thompson, Dallas E Peterson, Jeroen Roelofs, Mithila Jugulam
ABSTRACT
Herbicides that inhibit hydroxyphenylpyruvate dioxygenase (HPPD) such as mesotrione are widely used to control a broad spectrum of weeds in agriculture.