Merck
CD14
Hepatology (Baltimore, Md.) (2016-09-20)
Akira Nishio, Tomohide Tatsumi, Takatoshi Nawa, Takahiro Suda, Teppei Yoshioka, Yoshiki Onishi, Satoshi Aono, Minoru Shigekawa, Hayato Hikita, Ryotaro Sakamori, Daisuke Okuzaki, Takasuke Fukuhara, Yoshiharu Matsuura, Naoki Hiramatsu, Tetsuo Takehara
ABSTRACT
Natural killer (NK) cell activation is associated with both liver injury and persistent infection in chronic hepatitis C (CHC); however, the detailed mechanism of this activation has not yet been fully elucidated. Because galectin-9 (Gal-9) has been reported to be increased in the serum and liver tissue of CHC patients, we investigated the function of Gal-9 in NK cell activation in CHC. First, we evaluated the function of Gal-9 on NK cytotoxicity in vitro. Gal-9 treatment resulted in increased cytotoxicity of naïve NK cells, and the Gal-9-activated NK cells demonstrated cytotoxicity toward hepatoma cells and T cells. Additionally, coculturing peripheral blood mononuclear cells (PBMCs) with JFH-1/Huh7.5.1 cells increased both Gal-9 production and NK cell cytotoxicity. Next, we investigated the source of Gal-9 and the mechanism of Gal-9 production. Deletion of CD14 These results demonstrate that CD14