Merck
  • Aerosolizable gold nano-in-micro dry powder formulations for theragnosis and lung delivery.
Aerosolizable gold nano-in-micro dry powder formulations for theragnosis and lung delivery.
International journal of pharmaceutics (2017-01-24)
A Sofia Silva, Ana M Sousa, Renato P Cabral, Marta C Silva, Clarinda Costa, Sónia P Miguel, Vasco D B Bonifácio, Teresa Casimiro, Ilídio J Correia, Ana Aguiar-Ricardo
ABSTRACT
Functionalized gold nanoparticles (AuNPs) have been widely investigated as promising multifunctional nanosystems for the theragnosis of lung cancer, the most common and prominent cause of cancer death worldwide. Nevertheless, nanoparticles are not in appropriate sizes for an accurate deep lung delivery and the lack of locally and effective delivery of therapeutic biomolecules to the deep lungs is, in fact, the major cause of low therapeutic outcome. Herein we incorporate, for the first time, AuNPs into respirable microparticles. AuNPs were functionalized with biocompatible oligo(2-oxazoline)-based optically stable fluorescent coatings, and conjugated with a laminin peptide (YIGSR) for targeted lung cancer delivery. These POxylated AuNPs were then incorporated into a chitosan matrix by a clean process, supercritical CO