Merck
  • A hydantoin isoform of cyclic N6-threonylcarbamoyladenosine (ct6A) is present in tRNAs.
A hydantoin isoform of cyclic N6-threonylcarbamoyladenosine (ct6A) is present in tRNAs.
Nucleic acids research (2016-12-04)
Michal Matuszewski, Jakub Wojciechowski, Kenjyo Miyauchi, Zofia Gdaniec, Wojciech M Wolf, Tsutomu Suzuki, Elzbieta Sochacka
ABSTRACT
N6-Threonylcarbamoyladenosine (t6A) and its derivatives are universally conserved modified nucleosides found at position 37, 3΄ adjacent to the anticodon in tRNAs responsible for ANN codons. These modifications have pleiotropic functions of tRNAs in decoding and protein synthesis. In certain species of bacteria, fungi, plants and protists, t6A is further modified to the cyclic t6A (ct6A) via dehydration catalyzed by TcdA. This additional modification is involved in efficient decoding of tRNALys. Previous work indicated that the chemical structure of ct6A is a cyclic active ester with an oxazolone ring. In this study, we solved the crystal structure of chemically synthesized ct6A nucleoside. Unexpectedly, we found that the ct6A adopted a hydantoin isoform rather than an oxazolone isoform, and further showed that the hydantoin isoform of ct6A was actually present in Escherichia coli tRNAs. In addition, we observed that hydantoin ct6A is susceptible to epimerization under mild alkaline conditions, warning us to avoid conventional deacylation of tRNAs. A hallmark structural feature of this isoform is the twisted arrangement of the hydantoin and adenine rings. Functional roles of ct6A37 in tRNAs should be reconsidered.