Merck
  • TRAIL facilitates cytokine expression and macrophage migration during hypoxia/reoxygenation via ER stress-dependent NF-κB pathway.
TRAIL facilitates cytokine expression and macrophage migration during hypoxia/reoxygenation via ER stress-dependent NF-κB pathway.
Molecular immunology (2017-01-11)
Yinan Jiang, Xiaoyan Chen, Mengya Fan, Hui Li, Weina Zhu, Xi Chen, Chenghua Cao, Rui Xu, Yaohui Wang, Yuanfang Ma
ABSTRACT
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL), which is known as a key molecule to induce cancer cell apoptosis, has also been found to participate in the process of ischemia/reperfusion (I/R) injury. Infiltrated macrophages play dual roles in inflammatory injury and healing following I/R. Whether TRAIL has any effect on macrophages during this process remains elusive. Here we showed that I/R triggered the expressions of TRAIL, DR5 and cytokines (IL-1β, TNFα, CCL-2 and ICAM-1), in addition to macrophage infiltration, which could be abolished by TRAIL neutralizing antibody. In vitro, TRAIL enhanced DR5 expression and facilitated the macrophages migration following hypoxia/reoxygenation (H/R) treatment in a dose-dependent manner via ER stress and NF-κB signaling pathways, which is accompanied by inflammatory factors expression. The increased cytokines production (such as TNFα and IL-1β) stimulated by TRAIL can be blocked by the NF-κB and ER stress inhibitor. The results also suggested that NF-κB activation of macrophages during H/R was regulated by ER stress. Thus, our research present that TRAIL affects functional activities of macrophages during I/R injury, which may be a potential therapeutic target for ischemic heart disease.