Merck
  • Endoplasmic Reticulum Stress Induction and ERK1/2 Activation Contribute to Nefazodone-Induced Toxicity in Hepatic Cells.
Endoplasmic Reticulum Stress Induction and ERK1/2 Activation Contribute to Nefazodone-Induced Toxicity in Hepatic Cells.
Toxicological sciences : an official journal of the Society of Toxicology (2016-09-11)
Zhen Ren, Si Chen, Jie Zhang, Utkarsh Doshi, Albert P Li, Lei Guo
ABSTRACT
Nefazodone, an antagonist for the 5-hydroxytryptanine receptor, has been used for the treatment of depression. Acute liver injury has been documented to be associated with the use of nefazodone; however, the mechanisms of nefazodone-induced liver toxicity are not well defined. In this report, using biochemical and molecular analyses, we characterized the molecular mechanisms underlying the hepatotoxicity of nefazodone. We found that nefazodone induced endoplasmic reticulum (ER) stress in HepG2 cells, as the expression of typical ER stress markers, including CHOP, ATF-4, and p-eIF2α, was significantly increased, and splicing of XBP1 was observed. Nefazodone-suppressed protein secretion was evaluated using a Gaussia luciferase reporter assay that measures ER stress. The ER stress inhibitors (4-phenylbutyrate and salubrinal) and knockdown of ATF-4 gene attenuated nefazodone-induced ER stress and cytotoxicity. Nefazodone activated the MAPK signaling pathway, as indicated by increased phosphorylation of JNK, ERK1/2, and p38. Inhibition of ERK1/2 reduced ER stress caused by nefazodone. Taken together, our findings suggest that ER stress contributes to nefazodone-induced toxicity in HepG2 cells and that the MAPK signaling pathway plays an important role in ER stress.