- Disposition and metabolism of TAK-438 (vonoprazan fumarate), a novel potassium-competitive acid blocker, in rats and dogs.
Disposition and metabolism of TAK-438 (vonoprazan fumarate), a novel potassium-competitive acid blocker, in rats and dogs.
Xenobiotica; the fate of foreign compounds in biological systems (2016-05-27)
Akifumi Kogame, Toshiyuki Takeuchi, Masami Nonaka, Hitomi Yamasaki, Naohiro Kawaguchi, Ai Bernards, Yoshihiko Tagawa, Akio Morohashi, Takahiro Kondo, Toshiya Moriwaki, Satoru Asahi
PMID27225050
ABSTRACT
1. Following oral administration of [